Assay Details
Gene Symbol: MYC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142873.1 | NP_001136345.1 | 757 | 84 | ||
XM_015145930.1 | XP_015001416.1 | 1374 | 84 | |||
XM_028852119.1 | XP_028707952.1 | 1947 | 84 | |||
GenBank mRNA | FJ375753.1 | - | 757 | 84 | ||
JU475686.1 | - | 1319 | 84 | |||
JV047129.1 | - | 1214 | 84 | |||
JV636169.1 | - | 1214 | 84 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
