Assay Details
Gene Symbol: SELENOW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042737.2 | NP_001036202.1 | 5 - 6 | 473 | 100 | |
GenBank mRNA | JV047912.1 | - | 5 - 6 | 376 | 100 | |
JV625537.1 | - | 5 - 6 | 468 | 100 | ||
JV704030.1 | - | 5 - 6 | 473 | 101 | ||
U67450.1 | - | 5 - 6 | 331 | 100 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
