Assay Details
Gene Symbol: DYRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001086002.3 | XP_001086002.2 | 82 | 84 | ||
XM_001086117.4 | XP_001086117.2 | 82 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001086002.3, XM_001086117.4 transcripts.
Set Membership: |
![]() ![]() ![]() |
