Assay Details
Gene Symbol: CRADD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267004.1 | NP_001253933.1 | 96 | 101 | ||
XM_028828937.1 | XP_028684770.1 | 132 | 101 | |||
XM_028828938.1 | XP_028684771.1 | 132 | 101 | |||
XM_028828939.1 | XP_028684772.1 | 132 | 101 | |||
XM_028828940.1 | XP_028684773.1 | 132 | 101 | |||
XM_028828941.1 | XP_028684774.1 | 132 | 101 | |||
XM_028828942.1 | XP_028684775.1 | 132 | 101 | |||
XM_028828943.1 | XP_028684776.1 | 132 | 101 | |||
XM_028828944.1 | XP_028684777.1 | 132 | 101 | |||
XM_028828945.1 | XP_028684778.1 | 132 | 101 | |||
GenBank mRNA | JU336904.1 | - | 99 | 101 | ||
JU477647.1 | - | 96 | 101 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the NM_001267004.1, XM_028828937.1, XM_028828938.1, XM_028828939.1, XM_028828940.1, XM_028828941.1, XM_028828942.1, XM_028828943.1, XM_028828944.1, XM_028828945.1 transcripts.
This assay is in the 5'UTR region of the NM_001267004.1, XM_028828937.1, XM_028828938.1, XM_028828939.1, XM_028828940.1, XM_028828941.1, XM_028828942.1, XM_028828943.1, XM_028828944.1, XM_028828945.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
