Assay Details
Gene Symbol: AKAP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001102077.4 | XP_001102077.1 | 1665 | 127 | ||
GenBank mRNA | JU323375.1 | - | 1623 | 127 | ||
JU472135.1 | - | 1527 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
