Assay Details
Gene Symbol: KCNJ9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001117363.4 | XP_001117363.1 | 284 | 120 | ||
GenBank mRNA | JU324473.1 | - | 94 | 120 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001117363.4 transcripts.
Set Membership: |
![]() ![]() |
