Assay Details
Gene Symbol: H1FX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266083.2 | NP_001253012.1 | 1 | 960 | 159 | |
GenBank mRNA | JU336969.1 | - | 1 | 990 | 159 | |
JV048617.1 | - | 1 | 960 | 159 | ||
JV486047.1 | - | 1 | 298 | 159 | ||
JV601127.1 | - | 1 | 1208 | 159 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
