Assay Details
Gene Symbol: TRH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015130168.2 | XP_014985654.2 | 133 | 135 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015130168.2 transcripts.
Set Membership: |
![]() ![]() |
