Assay Details
Gene Symbol: RCAN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015131310.2 | XP_014986796.1 | 536 | 152 | ||
XM_015131315.2 | XP_014986801.1 | 569 | 152 | |||
GenBank mRNA | JU321003.1 | - | 1 - 2 | 260 | 152 | |
JU472768.1 | - | 1 - 2 | 247 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
