Assay Details
Gene Symbol: SH3BGRL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015131943.2 | XP_014987429.1 | 1162 | 83 | ||
GenBank mRNA | JU320436.1 | - | 587 | 83 | ||
JU477415.1 | - | 378 | 83 | |||
JV045282.1 | - | 229 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
