Assay Details
Gene Symbol: LOC100429346 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015119784.2 | XP_014975270.1 | 1309 | 110 | ||
XM_015119785.2 | XP_014975271.1 | 1275 | 110 | |||
GenBank mRNA | JU320332.1 | - | 1042 | 110 | ||
JU320333.1 | - | 1240 | 110 | |||
JU320334.1 | - | 1240 | 110 | |||
JU321739.1 | - | 1240 | 110 | |||
JU471733.1 | - | 1056 | 110 | |||
JV044188.1 | - | 1254 | 110 | |||
JV046772.1 | - | 1254 | 110 |
Gene Symbol: TUBG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261011.1 | NP_001247940.1 | 1254 | 110 | ||
GenBank mRNA | JU471734.1 | - | 1056 | 110 | ||
JV044189.1 | - | 1254 | 110 | |||
JV635408.1 | - | 1058 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
