Assay Details
Gene Symbol: ARPP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266449.1 | NP_001253378.1 | 120 | 101 | ||
GenBank mRNA | JU332598.1 | - | 128 | 101 | ||
JU332599.1 | - | 128 | 101 | |||
JU332600.1 | - | 128 | 101 | |||
JU332601.1 | - | 128 | 101 | |||
JU332602.1 | - | 128 | 101 | |||
JU332603.1 | - | 128 | 101 | |||
JU332604.1 | - | 128 | 101 | |||
JU332605.1 | - | 128 | 101 | |||
JV045981.1 | - | 120 | 101 | |||
JV045982.1 | - | 120 | 101 | |||
JV045983.1 | - | 120 | 101 | |||
JV045984.1 | - | 120 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001266449.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
