Assay Details
Gene Symbol: EHD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257730.1 | NP_001244659.1 | 2 - 3 | 1001 | 165 | |
GenBank mRNA | JU321915.1 | - | 2 - 3 | 1001 | 165 | |
JU321916.1 | - | 2 - 3 | 1001 | 165 | ||
JV600498.1 | - | 2 - 3 | 1001 | 165 | ||
JV668205.1 | - | 2 - 3 | 980 | 165 |

More Information
Set Membership: |
![]() ![]() ![]() |
