Assay Details
Gene Symbol: MRPL20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260920.1 | NP_001247849.1 | 154 | 114 | ||
GenBank mRNA | JU322855.1 | - | 166 | 114 | ||
JU472232.1 | - | 162 | 114 | |||
JV044834.1 | - | 154 | 114 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
