Assay Details
Gene Symbol: MRPS36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258095.1 | NP_001245024.1 | 1 - 2 | 116 | 133 | |
GenBank mRNA | JU471737.1 | - | 1 - 2 | 116 | 133 | |
JV044394.1 | - | 1 - 2 | 115 | 133 | ||
JV602488.1 | - | 1 - 2 | 117 | 133 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001258095.1 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
