Assay Details
Gene Symbol: GGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015149917.2 | XP_015005403.2 | 3919 | 85 | ||
XM_015149918.2 | XP_015005404.2 | 4039 | 85 | |||
XM_015149919.2 | XP_015005405.2 | 2107 | 85 | |||
XM_015149920.2 | XP_015005406.2 | 2158 | 85 | |||
XM_015149921.2 | XP_015005407.2 | 1969 | 85 | |||
XM_015149922.2 | XP_015005408.2 | 2094 | 85 | |||
XM_015149924.2 | XP_015005410.2 | 3928 | 85 | |||
XM_015149925.2 | XP_015005411.2 | 2122 | 85 | |||
XM_015149926.2 | XP_015005412.2 | 2228 | 85 | |||
XM_015149927.2 | XP_015005413.2 | 2110 | 85 | |||
XM_015149928.2 | XP_015005414.2 | 3917 | 85 | |||
XM_015149929.2 | XP_015005415.2 | 3931 | 85 | |||
XM_015149930.2 | XP_015005416.2 | 1935 | 85 | |||
XM_015149931.2 | XP_015005417.2 | 3926 | 85 | |||
XM_015149932.2 | XP_015005418.2 | 1996 | 85 | |||
XM_015149936.2 | XP_015005422.1 | 812 | 85 | |||
XM_028828623.1 | XP_028684456.1 | 4036 | 85 | |||
XM_028828624.1 | XP_028684457.1 | 2225 | 85 | |||
XM_028828626.1 | XP_028684459.1 | 1970 | 85 | |||
XM_028828627.1 | XP_028684460.1 | 4034 | 85 | |||
XM_028828628.1 | XP_028684461.1 | 2161 | 85 |
Gene Symbol: LOC700936 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028828571.1 | XP_028684404.1 | 1627 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
