Assay Details
Gene Symbol: DAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015130798.2 | XP_014986284.1 | 89 | 102 | ||
XM_015130799.2 | XP_014986285.1 | 48 | 102 | |||
XM_015130807.2 | XP_014986293.1 | 48 | 102 | |||
XM_015130809.2 | XP_014986295.1 | 48 | 102 | |||
XM_015130810.2 | XP_014986296.1 | 48 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015130798.2, XM_015130799.2, XM_015130807.2, XM_015130809.2, XM_015130810.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
