Assay Details
Gene Symbol: SUMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195642.1 | NP_001182571.1 | 49 | 136 | ||
GenBank mRNA | CD766851.1 | - | 52 | 136 | ||
JU319857.1 | - | 39 | 136 | |||
JU336217.1 | - | 39 | 136 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001195642.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
