Assay Details
Gene Symbol: LPAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260860.1 | NP_001247789.1 | 1 | 46 | 150 | |
GenBank mRNA | JU320067.1 | - | 1 | 46 | 150 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001260860.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
