Assay Details
Gene Symbol: PFN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015118359.2 | XP_014973845.1 | 711 | 116 | ||
GenBank mRNA | JU320596.1 | - | 1 | 120 | 116 | |
JU320597.1 | - | 1 | 120 | 116 | ||
JV043879.1 | - | 1 | 155 | 116 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015118359.2 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
