Assay Details
Gene Symbol: RBBP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260771.1 | NP_001247700.1 | 72 | 86 | ||
GenBank mRNA | JU332304.1 | - | 72 | 86 | ||
JU332305.1 | - | 72 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001260771.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
