Assay Details
Gene Symbol: CDKN2AIPNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015140929.2 | XP_014996415.1 | 93 | 150 | ||
XM_015140930.2 | XP_014996416.1 | 91 | 150 | |||
GenBank mRNA | JU337052.1 | - | 47 | 150 | ||
JU477780.1 | - | 45 | 150 | |||
JV598021.1 | - | 53 | 150 | |||
JV598023.1 | - | 53 | 150 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015140929.2, XM_015140930.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
