Assay Details
Gene Symbol: PML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042434.1 | NP_001035899.1 | 131 | 117 | ||
XM_015142614.2 | XP_014998100.1 | 266 | 117 | |||
XM_015142615.2 | XP_014998101.1 | 266 | 117 | |||
XM_015142616.2 | XP_014998102.1 | 266 | 117 | |||
XM_015142617.2 | XP_014998103.1 | 266 | 117 | |||
XM_015142618.2 | XP_014998104.1 | 266 | 117 | |||
XM_015142619.2 | XP_014998105.1 | 266 | 117 | |||
XM_015142620.2 | XP_014998106.1 | 266 | 117 | |||
XM_015142621.2 | XP_014998107.1 | 266 | 117 | |||
XM_015142622.2 | XP_014998108.1 | 266 | 117 | |||
XM_028850268.1 | XP_028706101.1 | 266 | 117 | |||
XM_028850269.1 | XP_028706102.1 | 266 | 117 | |||
XM_028850270.1 | XP_028706103.1 | 266 | 117 | |||
XM_028850271.1 | XP_028706104.1 | 266 | 117 | |||
GenBank mRNA | DQ231471.1 | - | 131 | 117 | ||
JU323536.1 | - | 253 | 117 | |||
JU323852.1 | - | 253 | 117 | |||
JU476034.1 | - | 215 | 117 | |||
JU476035.1 | - | 215 | 117 | |||
JV635596.1 | - | 152 | 117 |

More Information
Set Membership: |
![]() ![]() |
