Assay Details
Gene Symbol: PML | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042434.1 | NP_001035899.1 | 1859 | 114 | ||
XM_015142614.2 | XP_014998100.1 | 1850 | 114 | |||
GenBank mRNA | DQ231471.1 | - | 1859 | 114 | ||
JU323536.1 | - | 1837 | 114 | |||
JU323852.1 | - | 1981 | 114 | |||
JU476034.1 | - | 1799 | 114 | |||
JU476035.1 | - | 1943 | 114 | |||
JV635596.1 | - | 1880 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() |
