Assay Details
Gene Symbol: ACTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033084.1 | NP_001028256.1 | 1000 | 90 | ||
GenBank mRNA | AY369786.1 | - | 219 | 90 | ||
JV480231.1 | - | 452 | 90 |
Gene Symbol: LOC705671 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028844381.1 | XP_028700214.1 | 924 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
