Assay Details
Gene Symbol: GHV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042739.1 | NP_001036204.1 | 499 | 152 | ||
XM_028835510.1 | XP_028691343.1 | 937 | 152 | |||
GenBank mRNA | L16555.1 | - | 499 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
