Assay Details
Gene Symbol: CHMP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085358.2 | NP_001078827.1 | 504 | 65 | ||
GenBank mRNA | JU487729.1 | - | 495 | 65 | ||
JV618545.1 | - | 499 | 65 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
