Assay Details
Gene Symbol: DIO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_159356.2 | 2 - 3 | 377 | 82 | ||
GenBank mRNA | JU332318.1 | - | 2 - 3 | 377 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
