Assay Details
Gene Symbol: GPX7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159368.1 | NP_001152840.1 | 1 - 2 | 165 | 90 | |
GenBank mRNA | JU326961.1 | - | 1 - 2 | 71 | 90 | |
JU485045.1 | - | 1 - 2 | 122 | 90 | ||
JV058866.1 | - | 1 - 2 | 139 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
