Assay Details
Gene Symbol: SELENOF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159416.2 | NP_001152888.1 | 630 | 98 | ||
NR_144533.2 | 380 | 98 | ||||
GenBank mRNA | JU332145.1 | - | 395 | 98 | ||
JV528708.1 | - | 404 | 98 | |||
JV704574.1 | - | 386 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
