Assay Details
Gene Symbol: MIR128A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_031965.1 | 1 | 38 | 74 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc finger RANBP2-type containing 3 | ZRANB3 | -136.3kb | Anti-sense | Chr. 12 - 20372735 - 20680125 on NCBI Build Mmul_10 | |
Downstream | uncharacterized LOC114671317 | LOC114671317 | 75.4kb | Anti-sense | Chr. 12 - 20891910 - 20894428 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
