Assay Details
Gene Symbol: MIR455 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032534.1 | 1 | 48 | 67 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | tight junction protein 2 | TJP2 | Sense | Chr. 15 - 84575055 - 84714613 on NCBI Build Mmul_10 | ||
Downstream | family with sequence similarity 189 member A2 | FAM189A2 | C15H9orf61 | 134.5kb | Sense | Chr. 15 - 84774221 - 84839033 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
