Assay Details
Gene Symbol: RELL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193775.2 | NP_001180704.1 | 3 - 4 | 467 | 65 | |
XM_015138139.2 | XP_014993625.1 | 500 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
