Assay Details
Gene Symbol: METTL24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193830.2 | NP_001180759.1 | 2 - 3 | 418 | 74 | |
XM_028846658.1 | XP_028702491.1 | 545 | 74 | |||
GenBank mRNA | JU330569.1 | - | 1 - 2 | 79 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
