Assay Details
Gene Symbol: METTL24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193830.2 | NP_001180759.1 | 4 - 5 | 783 | 87 | |
XM_028846658.1 | XP_028702491.1 | 910 | 87 | |||
GenBank mRNA | JU330569.1 | - | 3 - 4 | 444 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
