Assay Details
Gene Symbol: SMIM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194105.1 | NP_001181034.1 | 38 | 54 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194105.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
