Assay Details
Gene Symbol: HSCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194228.1 | NP_001181157.1 | 614 | 82 | ||
XM_028827360.1 | XP_028683193.1 | 649 | 82 | |||
XM_028827364.1 | XP_028683197.1 | 592 | 82 | |||
XM_028827365.1 | XP_028683198.1 | 334 | 82 | |||
GenBank mRNA | JU325710.1 | - | 549 | 82 | ||
JV046269.1 | - | 583 | 82 | |||
JV635541.1 | - | 569 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
