Assay Details
Gene Symbol: C20H16orf72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194229.2 | NP_001181158.1 | 376 | 81 | ||
GenBank mRNA | JU319631.1 | - | 329 | 81 | ||
JU475279.1 | - | 522 | 81 | |||
JV504087.1 | - | 330 | 81 | |||
JV579513.1 | - | 353 | 81 | |||
JV601007.1 | - | 632 | 81 |
