Assay Details
Gene Symbol: BLOC1S5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194862.4 | NP_001181791.1 | 3 - 4 | 372 | 76 | |
XM_015135443.2 | XP_014990929.1 | 383 | 76 | |||
GenBank mRNA | JU323026.1 | - | 3 - 4 | 340 | 76 | |
JV045459.1 | - | 3 - 4 | 350 | 76 | ||
JV658461.1 | - | 3 - 4 | 353 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
