Assay Details
Gene Symbol: BLOC1S5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194862.4 | NP_001181791.1 | 431 | 69 | ||
XM_015135443.2 | XP_014990929.1 | 442 | 69 | |||
XM_015135444.2 | XP_014990930.1 | 306 | 69 | |||
GenBank mRNA | JU323026.1 | - | 399 | 69 | ||
JV045459.1 | - | 409 | 69 | |||
JV658461.1 | - | 412 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
