Assay Details
Gene Symbol: PLA2G12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194877.3 | NP_001181806.1 | 713 | 82 | ||
GenBank mRNA | JU321069.1 | - | 531 | 82 | ||
JU332210.1 | - | 531 | 82 | |||
JU332211.1 | - | 531 | 82 | |||
JU332212.1 | - | 531 | 82 | |||
JU473084.1 | - | 481 | 82 | |||
JV636633.1 | - | 633 | 82 |
Gene Symbol: DCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV046313.1 | - | 526 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
