Assay Details
Gene Symbol: CXHXorf65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190834.1 | NP_001177763.1 | 426 | 89 | ||
GenBank mRNA | JU481289.1 | - | 643 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
