Assay Details
Gene Symbol: COMMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195447.1 | NP_001182376.1 | 538 | 98 | ||
XM_028853230.1 | XP_028709063.1 | 667 | 98 | |||
GenBank mRNA | CO726581.1 | - | 542 | 98 | ||
DR770072.1 | - | 460 | 98 | |||
JU333684.1 | - | 547 | 98 | |||
JU474933.1 | - | 575 | 98 | |||
JV043494.1 | - | 547 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
