Assay Details
Gene Symbol: GNB1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190909.2 | NP_001177838.2 | 2 - 3 | 261 | 76 | |
XM_015149651.2 | XP_015005137.2 | 490 | 76 | |||
XM_028827449.1 | XP_028683282.1 | 693 | 76 | |||
XM_028827450.1 | XP_028683283.1 | 597 | 76 | |||
XM_028827451.1 | XP_028683284.1 | 541 | 76 | |||
GenBank mRNA | JU328988.1 | - | 1 - 2 | 53 | 76 | |
JU476095.1 | - | 4 - 5 | 562 | 76 | ||
JV717368.1 | - | 1 - 2 | 98 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
