Assay Details
Gene Symbol: UBA52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195756.1 | NP_001182685.1 | 39 | 75 | ||
XM_015123316.2 | XP_014978802.1 | 140 | 75 | |||
GenBank mRNA | DR770931.1 | - | 41 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001195756.1, XM_015123316.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
