Assay Details
Gene Symbol: FPGT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204799.2 | NP_001191728.2 | 3 - 4 | 409 | 100 | |
GenBank mRNA | JU323979.1 | - | 3 - 4 | 369 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
