Assay Details
Gene Symbol: RPLP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193576.1 | NP_001180505.1 | 41 | 76 | ||
GenBank mRNA | CO582752.1 | - | 58 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001193576.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
