Assay Details
Gene Symbol: GPR171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194365.1 | NP_001181294.1 | 80 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001194365.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
