Assay Details
Gene Symbol: FAM222B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001109457.4 | XP_001109457.3 | 158 | 77 | ||
XM_028835562.1 | XP_028691395.1 | 159 | 77 | |||
GenBank mRNA | JU335411.1 | - | 43 | 77 | ||
JU472967.1 | - | 50 | 77 | |||
JV704445.1 | - | 85 | 77 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001109457.4, XM_028835562.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
