Assay Details
Gene Symbol: FAM222B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015118848.2 | XP_014974334.2 | 318 | 94 | ||
XM_015118849.2 | XP_014974335.2 | 374 | 94 | |||
XM_028835562.1 | XP_028691395.1 | 284 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015118848.2, XM_015118849.2, XM_028835562.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
